2,2'-Bis(dicyclohexylphosphino)-1,1'-biphenyl 98%

Supplier: Ambeed
Warning

A700439-1G A700439-250MG
77255-176EA 265.74 USD
77255-176 77255-178
2,2'-Bis(dicyclohexylphosphino)-1,1'-biphenyl 98%
2,2'-Bis(dicyclohexylphosphino)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₃₆H₅₂P₂
MW: 546.7456 g/mol
MDL Number: MFCD07368358
CAS Number: 255897-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR