1-Bromo-4-chlorobutane 97%

Supplier: Ambeed
Danger

A699581-10G A699581-25G
77243-462EA 5.15 USD
77243-462 77243-464
1-Bromo-4-chlorobutane 97%
1-Bromo-4-chlorobutane

Precaución: for Research Use Only

Formula: BrH₂C(CH₂)₂CH₂Cl
MW: 171.46 g/mol
MDL Number: MFCD00001010
CAS Number: 6940-78-9
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR