1-Cyclohexyl-3-phenylthiourea 97%

Supplier: Ambeed
Danger

A699571-1G A699571-25G A699571-5G
77243-456EA 10 USD
77243-456 77243-458 77243-460
1-Cyclohexyl-3-phenylthiourea 97%
1-Cyclohexyl-3-phenylthiourea

Precaución: for Research Use Only

Formula: C₁₃H₁₈N₂S
MDL Number: MFCD00021317
CAS Number: 722-03-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR