2-[Bis(3,5-dimethylphenyl)phosphino]benzaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 2-[Bis(3,5-dimethylphenyl)phosphanyl]benzaldehyde, 0, 2-(Bis(3,5-dimethylphenyl)phosphino)benzaldehyde

A699081-100MG A699081-1G A699081-250MG
77255-160EA 49.44 USD
77255-160 77255-162 77255-164
2-[Bis(3,5-dimethylphenyl)phosphino]benzaldehyde 98%
2-[Bis(3,5-dimethylphenyl)phosphino]benzaldehyde

Precaución: for Research Use Only

Formula: C₂₃H₂₃OP
MW: 346.41 g/mol
MDL Number: MFCD15143662
CAS Number: 669091-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR