2,3-Difluoro-4-methoxybenzoic acid 98%

Supplier: Ambeed
Warning

A698944-1G A698944-250MG
77255-156EA 103 USD
77255-156 77255-158
2,3-Difluoro-4-methoxybenzoic acid 98%
2,3-Difluoro-4-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆F₂O₃
MDL Number: MFCD03428569
CAS Number: 329014-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR