6-((Naphthalen-1-ylmethyl)thio)-7H-purine 99%

Supplier: Ambeed
Warning

Synonyms: PU02

A698789-10MG A698789-1MG A698789-50MG
77243-450EA 53 USD
77243-450 77243-452 77243-454
6-((Naphthalen-1-ylmethyl)thio)-7H-purine 99%
6-((Naphthalen-1-ylmethyl)thio)-7H-purine

Precaución: for Research Use Only

Formula: C₁₆H₁₂N₄S
MDL Number: MFCD23381174
CAS Number: 313984-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR