3-Chloro-2,6-difluorobenzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: (3-Chloro-2,6-difluorophenyl)methanol

A698561-1G A698561-25G A698561-5G
77280-822EA 19 USD
77280-822 77280-824 77280-826
3-Chloro-2,6-difluorobenzyl alcohol 98%
3-Chloro-2,6-difluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₅ClF₂O
MDL Number: MFCD01631325
CAS Number: 252004-35-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR