4,6-Dibromo-1H-pyrrolo[3,2-c]pyridine 97%

Supplier: Ambeed
Warning

A698297-100MG A698297-250MG
77267-572EA 155.53 USD
77267-572 77267-574
4,6-Dibromo-1H-pyrrolo[3,2-c]pyridine 97%
4,6-Dibromo-1H-pyrrolo[3,2-c]pyridine

Precaución: for Research Use Only

Formula: C₇H₄Br₂N₂
MW: 275.92 g/mol
CAS Number: 2089314-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR