4-Chloro-3-nitro-5-(trifluoromethyl)benzoic acid 96%

Supplier: Ambeed
Warning

A697896-100MG A697896-1G A697896-250MG A697896-5G
77293-260EA 5 USD
77293-260 77293-262 77293-264 77293-266
4-Chloro-3-nitro-5-(trifluoromethyl)benzoic acid 96%
4-Chloro-3-nitro-5-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₃ClF₃NO₄
MDL Number: MFCD04972737
CAS Number: 350488-79-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR