AM-095 98%

Supplier: Ambeed
Warning

A697231-10MG A697231-1MG A697231-25MG A697231-50MG A697231-5MG
77243-424EA 47 USD
77243-424 77243-426 77243-428 77243-430 77243-432
AM-095 98%
AM-095

Precaución: for Research Use Only

Formula: C₂₇H₂₄N₂O₅
MDL Number: MFCD22665734
CAS Number: 1228690-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR