2-(p-Tolyl)pyridine 97%

Supplier: Ambeed
Warning

A697109-100G A697109-10G A697109-1G A697109-25G A697109-5G
77255-134EA 168.92 USD
77255-134 77255-136 77255-138 77255-140 77255-142
2-(p-Tolyl)pyridine 97%
2-(p-Tolyl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₁N
MW: 169.22 g/mol
MDL Number: MFCD00006281
CAS Number: 4467-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR