1-Ethyl-3-methylimidazoliumthiocyanate 98%

Supplier: Ambeed
Danger

Synonyms: 1-Ethyl-3-methyl-1H-imidazol-3-ium thiocyanate

A696784-100G A696784-25G
77267-568EA 163.77 USD
77267-568 77267-570
1-Ethyl-3-methylimidazoliumthiocyanate 98%
1-Ethyl-3-methylimidazoliumthiocyanate

Precaución: for Research Use Only

Formula: C₇H₁₁N₃S
MW: 169.25 g/mol
MDL Number: MFCD06798182
CAS Number: 331717-63-6
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR