3,4-Hexanedione 96%

Supplier: Ambeed
Danger

A696767-25G A696767-5G
77280-814EA 16.48 USD
77280-814 77280-816
3,4-Hexanedione 96%
3,4-Hexanedione

Precaución: for Research Use Only

Formula: C₆H₁₀O₂
MW: 114.14 g/mol
MDL Number: MFCD00010237
CAS Number: 4437-51-8
UN: 1224
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR