4-Benzyloxyphenyl-4-hydroxyphenyl sulfone 98%

Supplier: Ambeed
Warning

A696622-100G A696622-25G
77293-250EA 25 USD
77293-250 77293-252
4-Benzyloxyphenyl-4-hydroxyphenyl sulfone 98%
4-Benzyloxyphenyl-4-hydroxyphenyl sulfone

Precaución: for Research Use Only

Formula: C₁₉H₁₆O₄S
MDL Number: MFCD00185925
CAS Number: 63134-33-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR