4-Fluoro-N-methylbenzenesulphonamide 98%

Supplier: Ambeed
Warning

A696388-1G A696388-250MG A696388-25G A696388-5G
77293-242EA 25 USD
77293-242 77293-244 77293-246 77293-248
4-Fluoro-N-methylbenzenesulphonamide 98%
4-Fluoro-N-methylbenzenesulphonamide

Precaución: for Research Use Only

Formula: C₇H₈FNO₂S
MDL Number: MFCD01211989
CAS Number: 433-14-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR