2-Ethylhexyl palmitate 98%

Supplier: Ambeed
Warning

A696330-100G A696330-25G A696330-5G
77267-550EA 79 USD
77267-550 77267-552 77267-554
2-Ethylhexyl palmitate 98%
2-Ethylhexyl palmitate

Precaución: for Research Use Only

Formula: C₂₄H₄₈O₂
MDL Number: MFCD00072255
CAS Number: 29806-73-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR