6-Fluoroveratraldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-4,5-dimethoxybenzaldehyde, 4,5-Dimethoxy-2-fluorobenzaldehyde, 3,4-Dimethoxy-6-fluorobenzaldehyde

A695997-10G A695997-5G
77280-810EA 219.39 USD
77280-810 77280-812
6-Fluoroveratraldehyde 98%
6-Fluoroveratraldehyde

Precaución: for Research Use Only

Formula: C₉H₉FO₃
MW: 184.17 g/mol
MDL Number: MFCD00061108
CAS Number: 71924-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR