3-Methyl-4-nitrobenzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: (3-Methyl-4-nitrophenyl)methanol, 5-(Hydroxymethyl)-2-nitrotoluene

A695992-100G A695992-1G A695992-25G A695992-5G
77255-120EA 664.35 USD
77255-120 77255-122 77255-124 77255-126
3-Methyl-4-nitrobenzyl alcohol 98%
3-Methyl-4-nitrobenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.16 g/mol
MDL Number: MFCD00007171
CAS Number: 80866-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR