2-Methoxy-5-nitrobenzene-1-sulfonyl chloride 95%

Supplier: Ambeed
Danger

A695338-100MG A695338-1G A695338-5G A695338-250MG
77267-538EA 35 USD
77267-538 77267-540 77267-542 77617-876
2-Methoxy-5-nitrobenzene-1-sulfonyl chloride 95%
2-Methoxy-5-nitrobenzene-1-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₇H₆ClNO₅S
MW: 251.64 g/mol
MDL Number: MFCD07343894
CAS Number: 81118-92-5
UN: 3261
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR