3-Amino-2,2-dimethylpropionamide 96%
Supplier: Ambeed
|
Warning
|
A695145-100G
A695145-10G
A695145-1G
A695145-25G
A695145-5G
77280-800EA
175
USD
77280-800
77280-802
77280-804
77280-806
77280-808
3-Amino-2,2-dimethylpropionamide 96%
3-Amino-2,2-dimethylpropionamide
Precaución: for Research Use Only
Formula:
C5H12N2O |
MDL Number:
MFCD06797568 CAS Number: 324763-51-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...