Dichloro(norbornadiene)platinum(II) 98%

Supplier: Ambeed
Warning

A695138-100MG A695138-250MG
77255-114EA 56.65 USD
77255-114 77255-116
Dichloro(norbornadiene)platinum(II) 98%
Dichloro(norbornadiene)platinum(II)

Precaución: for Research Use Only

Formula: C₇H₈Cl₂Pt
MW: 358.125 g/mol
MDL Number: MFCD00213423
CAS Number: 12152-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR