Diethyl 2-methylthiazole-4,5-dicarboxylate 97%
Supplier: Ambeed
|
Warning
|
A694976-1G
A694976-250MG
A694976-25G
77280-796EA
48
USD
77280-796
77280-798
77749-429
Diethyl 2-methylthiazole-4,5-dicarboxylate 97%
Diethyl 2-methylthiazole-4,5-dicarboxylate
Precaución: for Research Use Only
Formula:
C₁₀H₁₃NO₄S MW: 243.27 g/mol |
MDL Number:
MFCD03789561 CAS Number: 18903-17-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...