1-Ethyl-2-iodobenzene 97%

Supplier: Ambeed
Warning

Synonyms: 2-Ethyliodobenzene, 1-Iodo-2-ethylbenzene

A694079-5G A694079-25G A694079-100G
77267-512EA 6.18 USD
77267-512 77749-427 77749-426
1-Ethyl-2-iodobenzene 97%
1-Ethyl-2-iodobenzene

Precaución: for Research Use Only

Formula: C₈H₉I
MW: 232.06 g/mol
MDL Number: MFCD00040871
CAS Number: 18282-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR