5-Hydroxy-1H-benzo[d]imidazole-4-carboxylic acid 95%

Supplier: Ambeed
Warning

A693451-100MG A693451-1G A693451-250MG
77243-396EA 150.38 USD
77243-396 77243-398 77243-400
5-Hydroxy-1H-benzo[d]imidazole-4-carboxylic acid 95%
5-Hydroxy-1H-benzo[d]imidazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₆N₂O₃
MW: 178.14 g/mol
MDL Number: MFCD22371733
CAS Number: 1378261-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR