4(7)-Nitrobenzimidazole 98%

Supplier: Ambeed
Warning

Synonyms: 7-Nitro-1H-benzo[d]imidazole

A693325-100MG A693325-1G A693325-250MG
77293-220EA 15 USD
77293-220 77293-222 77293-224
4(7)-Nitrobenzimidazole 98%
4(7)-Nitrobenzimidazole

Precaución: for Research Use Only

Formula: C₇H₅N₃O₂
MW: 163.134 g/mol
MDL Number: MFCD00220143
CAS Number: 10597-52-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR