5-Bromo-2-chloro-3-methoxybenzoic acid 97%

Supplier: Ambeed
Warning

A693187-1G A693187-250MG A693187-5G
77267-494EA 126 USD
77267-494 77267-496 77267-498
5-Bromo-2-chloro-3-methoxybenzoic acid 97%
5-Bromo-2-chloro-3-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆BrClO₃
MDL Number: MFCD29045016
CAS Number: 1415467-76-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR