1-Chloro-4-fluoro-2-iodobenzene 97%
Supplier: Ambeed
|
Warning
|
A692940-100G
A692940-25G
A692940-5G
77243-390EA
352.26
USD
77243-390
77243-392
77243-394
1-Chloro-4-fluoro-2-iodobenzene 97%
1-Chloro-4-fluoro-2-iodobenzene
Precaución: for Research Use Only
Formula:
C₆H₃ClFI MW: 256.4439 g/mol |
MDL Number:
MFCD00042575 CAS Number: 202982-68-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...