4-(Methylthio)benzoic acid 98%

Supplier: Ambeed
Warning

A692783-1G A692783-25G A692783-5G
77293-212EA 13 USD
77293-212 77293-214 77293-216
4-(Methylthio)benzoic acid 98%
4-(Methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₈O₂S
MDL Number: MFCD00002555
CAS Number: 13205-48-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR