3,5-Difluorobenzhydrazide 97%

Supplier: Ambeed
Warning

A692697-1G A692697-5G
77280-780EA 29 USD
77280-780 77280-782
3,5-Difluorobenzhydrazide 97%
3,5-Difluorobenzhydrazide

Precaución: for Research Use Only

Formula: C₇H₆F₂N₂O
MDL Number: MFCD01310810
CAS Number: 244022-63-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR