5-(4,6-Di((1R,5S)-3-oxa-8-azabicyclo[3.2.1]octan-8-yl)-1,3,5-triazin-2-yl)-4-(difluoromethyl)pyridin-2-amine 98+%

Supplier: Ambeed
Warning

Synonyms: PQR620

A692574-1MG A692574-5MG
77267-490EA 29.87 USD
77267-490 77267-492
5-(4,6-Di((1R,5S)-3-oxa-8-azabicyclo[3.2.1]octan-8-yl)-1,3,5-triazin-2-yl)-4-(difluoromethyl)pyridin-2-amine 98+%
5-(4,6-Di((1R,5S)-3-oxa-8-azabicyclo[3.2.1]octan-8-yl)-1,3,5-triazin-2-yl)-4-(difluoromethyl)pyridin-2-amine

Precaución: for Research Use Only

Formula: C₂₁H₂₅F₂N₇O₂
MW: 445.47 g/mol
MDL Number: MFCD30489733
CAS Number: 1927857-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR