1,2-Bis(chlorodimethylsilyl)ethane 95% (by GC)

Supplier: Ambeed
Danger

A692518-1G A692518-25G A692518-5G
77293-206EA 5.15 USD
77293-206 77293-208 77293-210
1,2-Bis(chlorodimethylsilyl)ethane 95% (by GC)
1,2-Bis(chlorodimethylsilyl)ethane

Precaución: for Research Use Only

Formula: C₆H₁₆Cl₂Si₂
MW: 215.26 g/mol
MDL Number: MFCD00000505
CAS Number: 13528-93-3
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR