2-Methyltetrahydrofuran-3-thiol (cis- and trans- mixture) 98%

Supplier: Ambeed
Danger

A692358-25G A692358-5G
77280-768EA 30 USD
77280-768 77280-770
2-Methyltetrahydrofuran-3-thiol (cis- and trans- mixture) 98%
2-Methyltetrahydrofuran-3-thiol (cis- and trans- mixture)

Precaución: for Research Use Only

Formula: C₅H₁₀OS
MDL Number: MFCD01632596
CAS Number: 57124-87-5
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR