3-(4-Chlorophenyl)propylamine 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-(4-Chlorophenyl)propan-1-amine
A692309-100MG
A692309-1G
A692309-250MG
77280-764EA
30.9
USD
77280-764
77280-766
77749-423
3-(4-Chlorophenyl)propylamine 95%
3-(4-Chlorophenyl)propylamine
Precaución: for Research Use Only
Formula:
C₉H₁₂ClN MW: 169.66 g/mol |
MDL Number:
MFCD06212968 CAS Number: 18655-50-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...