2-Bromo-5-iodo-4-nitro-1H-imidazole 97%

Supplier: Ambeed
Warning

A691604-100MG A691604-1G A691604-250MG A691604-5G
77267-478EA 32.96 USD
77267-478 77267-480 77267-482 77267-484
2-Bromo-5-iodo-4-nitro-1H-imidazole 97%
2-Bromo-5-iodo-4-nitro-1H-imidazole

Precaución: for Research Use Only

Formula: C₃HBrIN₃O₂
MW: 317.87 g/mol
MDL Number: MFCD10687146
CAS Number: 862895-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR