4-Methyl-2,1,3-benzothiadiazole 98%

Supplier: Ambeed
Warning

Synonyms: 4-Methylbenzo[c][1,2,5]thiadiazole

A691339-250MG A691339-1G A691339-5G
77293-204EA 33.99 USD
77293-204 77749-421 77749-422
4-Methyl-2,1,3-benzothiadiazole 98%
4-Methyl-2,1,3-benzothiadiazole

Precaución: for Research Use Only

Formula: C₇H₆N₂S
MW: 150.2009 g/mol
MDL Number: MFCD00126471
CAS Number: 1457-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR