2,2-Dichlorocyclopropylbenzene 97%

Supplier: Ambeed
Warning

A690749-5G A690749-25G
77267-466EA 17 USD
77267-466 77749-420
2,2-Dichlorocyclopropylbenzene 97%
2,2-Dichlorocyclopropylbenzene

Precaución: for Research Use Only

Formula: C₉H₈Cl₂
MDL Number: MFCD00001272
CAS Number: 2415-80-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR