2-Bromo-4-chloro-6-methylaniline 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-4-chloro-6-ethylaniline

A690541-100G A690541-1G A690541-25G A690541-5G
77267-458EA 372.86 USD
77267-458 77267-460 77267-462 77267-464
2-Bromo-4-chloro-6-methylaniline 98%
2-Bromo-4-chloro-6-methylaniline

Precaución: for Research Use Only

Formula: C₇H₇BrClN
MW: 220.4942 g/mol
MDL Number: MFCD07780637
CAS Number: 146948-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR