4-Benzyloxypyridine-N-oxide 98%

Supplier: Ambeed
Warning

A690518-1G A690518-25G A690518-5G
77293-188EA 17 USD
77293-188 77293-190 77293-192
4-Benzyloxypyridine-N-oxide 98%
4-Benzyloxypyridine-N-oxide

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO₂
MDL Number: MFCD00047427
CAS Number: 2683-66-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR