2-Hydroxy-3-nitrobenzonitrile 97%

Supplier: Ambeed
Warning

A690455-100MG A690455-250MG A690455-5G A690455-1G
77267-452EA 30.9 USD
77267-452 77267-454 77267-456 77749-419
2-Hydroxy-3-nitrobenzonitrile 97%
2-Hydroxy-3-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄N₂O₃
MW: 164.1200 g/mol
MDL Number: MFCD00234257
CAS Number: 28177-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR