3-(1-Hydroxyethyl)pyridine 98%

Supplier: Ambeed
Warning

Synonyms: 1-(Pyridin-3-yl)ethan-1-ol, 1-Hydroxy-1-(pyridin-3-yl)ethane

A689754-1G A689754-25G A689754-5G
77243-368EA 23.69 USD
77243-368 77243-370 77243-372
3-(1-Hydroxyethyl)pyridine 98%
3-(1-Hydroxyethyl)pyridine

Precaución: for Research Use Only

Formula: C₇H₉NO
MW: 123.152 g/mol
MDL Number: MFCD01691686
CAS Number: 4754-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR