2-Bromo-5-methylbenzotrifluoride 98%

Supplier: Ambeed
Danger

A689110-1G A689110-5G
77267-450EA 19.57 USD
77267-450 77747-385
2-Bromo-5-methylbenzotrifluoride 98%
2-Bromo-5-methylbenzotrifluoride

Precaución: for Research Use Only

Formula: C₈H₆BrF₃
MW: 239.04 g/mol
MDL Number: MFCD01631586
CAS Number: 261952-20-9
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR