2-(2,5-Dimethylphenyl)ethanol 95%

Supplier: Ambeed
Warning

A688338-10G A688338-25G A688338-5G
77255-076EA 91.67 USD
77255-076 77255-078 77255-080
2-(2,5-Dimethylphenyl)ethanol 95%
2-(2,5-Dimethylphenyl)ethanol

Precaución: for Research Use Only

Formula: C₁₀H₁₄O
MW: 150.218 g/mol
MDL Number: MFCD00020610
CAS Number: 6972-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR