2-(Trifluoromethyl)phenylacetylene 97%

Supplier: Ambeed
Danger

Synonyms: 1-Ethynyl-2-(trifluoromethyl)benzene

A688268-1G A688268-25G A688268-5G
77267-438EA 18.54 USD
77267-438 77267-440 77267-442
2-(Trifluoromethyl)phenylacetylene 97%
2-(Trifluoromethyl)phenylacetylene

Precaución: for Research Use Only

Formula: C₉H₅F₃
MW: 170.13 g/mol
MDL Number: MFCD00467313
CAS Number: 704-41-6
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR