(2-Bromo-4-methoxyphenyl)acetic acid 98%

Supplier: Ambeed
Warning

A687321-10G A687321-1G A687321-25G
77267-428EA 60 USD
77267-428 77267-430 77267-432
(2-Bromo-4-methoxyphenyl)acetic acid 98%
(2-Bromo-4-methoxyphenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
MDL Number: MFCD02664700
CAS Number: 66916-99-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR