3-(2-Chloropyrimidin-4-yl)benzoic acid 97%

Supplier: Ambeed
Warning

A686929-100MG A686929-1G A686929-250MG
77243-360EA 8.24 USD
77243-360 77243-362 77243-364
3-(2-Chloropyrimidin-4-yl)benzoic acid 97%
3-(2-Chloropyrimidin-4-yl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₇ClN₂O₂
MW: 234.64 g/mol
MDL Number: MFCD22628313
CAS Number: 937271-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR