Methyl-3,4-dihydroxyphenylacetate 98%

Supplier: Ambeed
Warning

Synonyms: 3,4-Dihydroxyphenylacetic Acid Methyl Ester, Methyl 2-(3,4-dihydroxyphenyl)acetate, Methyl 3,4-dihydroxyphenylacetate, Catechol-4-acetic Acid Methyl Ester, Homoprotocatechuic Acid Methyl Ester, Pyrocatechol-4-acetic Acid Methyl Ester

A686869-1G A686869-5G
77255-050EA 42.23 USD
77255-050 77255-052
Methyl-3,4-dihydroxyphenylacetate 98%
Methyl-3,4-dihydroxyphenylacetate

Precaución: for Research Use Only

Formula: C₉H₁₀O₄
MW: 182.18 g/mol
MDL Number: MFCD00583539
CAS Number: 25379-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR