5-Bromo-2,4-dimethoxybenzaldehyde 98%

Supplier: Ambeed
Warning

A686755-1G A686755-250MG A686755-5G
77267-422EA 33 USD
77267-422 77267-424 77267-426
5-Bromo-2,4-dimethoxybenzaldehyde 98%
5-Bromo-2,4-dimethoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
MDL Number: MFCD00010863
CAS Number: 130333-46-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR