(S)-2,2'-Bis(dicyclohexylphosphino)-1,1'-binaphthalene 98%

Supplier: Ambeed
Warning

A686467-100MG A686467-1G A686467-250MG
77267-410EA 151.41 USD
77267-410 77267-412 77267-414
(S)-2,2'-Bis(dicyclohexylphosphino)-1,1'-binaphthalene 98%
(S)-2,2'-Bis(dicyclohexylphosphino)-1,1'-binaphthalene

Precaución: for Research Use Only

Formula: C₄₄H₅₆P₂
MW: 646.86 g/mol
CAS Number: 121457-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR