1-Tetradecanol 98%

Supplier: Ambeed
Warning

Synonyms: 1-Tetradecyl alcohol, 1-Hydroxy tetradecane, Myristyl alcohol

A686448-100G A686448-25G A686448-500G
77243-354EA 8 USD
77243-354 77243-356 77243-358
1-Tetradecanol 98%
1-Tetradecanol

Precaución: for Research Use Only

Formula: H₃C(CH₂)₁₂CH₂OH
MW: 214.39 g/mol
Boiling Pt: 289
MDL Number: MFCD00004757
CAS Number: 112-72-1
Merck Index: 13,06361

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR