N-Iodosaccharin 98%

Supplier: Ambeed
Warning

A686404-1G A686404-250MG A686404-5G
77255-044EA 55 USD
77255-044 77255-046 77255-048
N-Iodosaccharin 98%
N-Iodosaccharin

Precaución: for Research Use Only

Formula: C₇H₄INO₃S
MDL Number: MFCD09037939
CAS Number: 86340-94-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR